ENA388699993 Enamine

(1S)-6-methoxy-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD15529805
  • Cas Number : 200425-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123389
Availability details for
Quantity
  • Availability
  • Add to Cart