ENA388699997 Enamine

5-sulfamoylisoquinoline-1-carboxylic acid

  • MDL number MFCD23143995
  • Cas Number : 1427380-39-9
  • Molecular Weight : 252.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-sulfamoylisoquinoline-1-carboxylic acid
Inchi Code 1S/C10H8N2O4S/c11-17(15,16)8-3-1-2-7-6(8)4-5-12-9(7)10(13)14/h1-5H,(H,13,14)(H2,11,15,16)
Inchi Key OVGSSIJBRCXYLD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119301

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-119301
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart