ENA388699999 Enamine

4-hydroxyoxane-4-carboxamide

  • MDL number MFCD16694136
  • Cas Number : 1250204-50-2
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxytetrahydro-2H-pyran-4-carboxamide
Inchi Code 1S/C6H11NO3/c7-5(8)6(9)1-3-10-4-2-6/h9H,1-4H2,(H2,7,8)
Inchi Key QQEWNVXINVYCDE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123402

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-123402
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart