ENA388700006 Enamine

(2,6-dichloro-4-fluorophenyl)hydrazine

  • MDL number MFCD19202959
  • Cas Number : 119453-09-7
  • Molecular Weight : 195.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-dichloro-4-fluorophenyl)hydrazine
Inchi Code 1S/C6H5Cl2FN2/c7-4-1-3(9)2-5(8)6(4)11-10/h1-2,11H,10H2
Inchi Key JASYLPFQMZCHAK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123413

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-123413
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart