ENA388700028 Enamine

10,13-diazatricyclo[7.4.1.0,5,14]tetradeca-5(14),6,8-trien-11-one

  • MDL number MFCD23143996
  • Cas Number : 1427379-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119392
Availability details for
Quantity
  • Availability
  • Add to Cart