ENA388700029 Enamine

dimethyl[3-(piperidin-4-yl)propyl]amine

  • MDL number MFCD16693192
  • Cas Number : 42270-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123458
Availability details for
Quantity
  • Availability
  • Add to Cart