ENA388700031 Enamine

6-({[(tert-butoxy)carbonyl]amino}methyl)-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD23144098
  • Cas Number : 1423471-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123460

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123460
Availability details for
Quantity
  • Availability
  • Add to Cart