ENA388700033 Enamine

3-(diphenylphosphanyl)benzene-1-sulfonic acid

  • MDL number MFCD06409941
  • Cas Number : 16704-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119475
Availability details for
Quantity
  • Availability
  • Add to Cart