ENA388700036 Enamine

{2-[(pyrrolidin-1-yl)methyl]phenyl}boronic acid

  • MDL number MFCD06801714
  • Cas Number : 878289-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123465
Availability details for
Quantity
  • Availability
  • Add to Cart