ENA388700041 Enamine

3-ethyl-2-propyl-1H-indole

  • MDL number MFCD21188793
  • Cas Number : 17901-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123472
Availability details for
Quantity
  • Availability
  • Add to Cart