ENA388700048 Enamine

1-(2-methylpropyl)-5-(propan-2-yl)-1H-pyrazol-4-amine

  • Molecular Weight : 181.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-isobutyl-5-isopropyl-1H-pyrazol-4-ylamine
Inchi Code 1S/C10H19N3/c1-7(2)6-13-10(8(3)4)9(11)5-12-13/h5,7-8H,6,11H2,1-4H3
Inchi Key IGAHNKCZXSNXIS-UHFFFAOYSA-N

Description

Physical Form liquid
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123479
Availability details for
Quantity
  • Availability
  • Add to Cart