ENA388700050 Enamine

9-bromo-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-2,5-dione

  • MDL number MFCD23143999
  • Cas Number : 1423723-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261...263
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119807
Availability details for
Quantity
  • Availability
  • Add to Cart