ENA388700057 Enamine

3,4-dimethyl-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD18892287
  • Cas Number : 1275222-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123523
Availability details for
Quantity
  • Availability
  • Add to Cart