ENA388700067 Enamine

2-(tert-butoxy)pyridine-4-carbonitrile

  • MDL number MFCD11196883
  • Cas Number : 1037131-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123542
Availability details for
Quantity
  • Availability
  • Add to Cart