ENA388700073 Enamine

6-hydroxyhexan-2-one

  • MDL number MFCD16165815
  • Cas Number : 21856-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123553
Availability details for
Quantity
  • Availability
  • Add to Cart