ENA388700080 Enamine

2-chloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}propan-1-one

  • MDL number MFCD11189414
  • Cas Number : 1037152-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123566
Availability details for
Quantity
  • Availability
  • Add to Cart