ENA388700083 Enamine

4-(1,2,3,6-tetrahydropyridin-4-yl)benzonitrile hydrochloride

  • MDL number MFCD23144110
  • Cas Number : 437998-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123569
Availability details for
Quantity
  • Availability
  • Add to Cart