ENA388700084 Enamine

3-phenylcycloheptan-1-ol

  • MDL number MFCD20341405
  • Cas Number : 19217-56-2
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-phenylcycloheptanol
Inchi Code 1S/C13H18O/c14-13-9-5-4-8-12(10-13)11-6-2-1-3-7-11/h1-3,6-7,12-14H,4-5,8-10H2
Inchi Key PWEFIUJVBSBVHN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123570
Availability details for
Quantity
  • Availability
  • Add to Cart