ENA388700086 Enamine

3-bromo-1-(4-fluorophenyl)propan-1-one

  • MDL number MFCD18391372
  • Cas Number : 57056-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107655
Availability details for
Quantity
  • Availability
  • Add to Cart