ENA388700091 Enamine

3-(2-bromophenyl)piperazin-2-one

  • MDL number MFCD07373588
  • Cas Number : 1246548-54-8
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-bromophenyl)-2-piperazinone
Inchi Code 1S/C10H11BrN2O/c11-8-4-2-1-3-7(8)9-10(14)13-6-5-12-9/h1-4,9,12H,5-6H2,(H,13,14)
Inchi Key MUQXZTVVAQKOSH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123586
Availability details for
Quantity
  • Availability
  • Add to Cart