ENA388700097 Enamine

2-(3-hydroxyphenoxy)-N-methylacetamide

  • MDL number MFCD11181968
  • Cas Number : 34918-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123592
Availability details for
Quantity
  • Availability
  • Add to Cart