ENA388700102 Enamine

2-[(oxiran-2-yl)methyl]-1,2-dihydrophthalazin-1-one

  • MDL number MFCD09730651
  • Cas Number : 953723-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123597
Availability details for
Quantity
  • Availability
  • Add to Cart