ENA388700105 Enamine

4-bromo-2,6-difluorobenzamide

  • MDL number MFCD13689151
  • Cas Number : 840481-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123602

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-123602
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart