ENA388700106 Enamine

2-(3-amino-5-methyl-1H-pyrazol-1-yl)-N-methylacetamide

  • MDL number MFCD15976670
  • Cas Number : 1379199-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123603

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123603
Availability details for
Quantity
  • Availability
  • Add to Cart