ENA388700107 Enamine

1-(3-fluoropyridin-2-yl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD16859680
  • Cas Number : 1342084-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123604
Availability details for
Quantity
  • Availability
  • Add to Cart