ENA388700110 Enamine

N-[2-(3-methylphenyl)ethyl]-3-(2-oxo-1,2-dihydroquinolin-3-yl)propanamide

  • MDL number MFCD16937234
  • Cas Number : 1328653-50-4
  • Molecular Weight : 334.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-hydroxy-3-quinolinyl)-N-[2-(3-methylphenyl)ethyl]propanamide
Inchi Code 1S/C21H22N2O2/c1-15-5-4-6-16(13-15)11-12-22-20(24)10-9-18-14-17-7-2-3-8-19(17)23-21(18)25/h2-8,13-14H,9-12H2,1H3,(H,22,24)(H,23,25)
Inchi Key SESBGWPELNSJLD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123609
Availability details for
Quantity
  • Availability
  • Add to Cart