ENA388700114 Enamine

2-amino-N-[(1,5-dimethyl-1H-pyrrol-2-yl)methyl]-2-methylpentanamide

  • MDL number MFCD12537429
  • Cas Number : 1182797-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123613
Availability details for
Quantity
  • Availability
  • Add to Cart