ENA388700120 Enamine

3-amino-4-methylpentanoic acid hydrochloride

  • MDL number MFCD18207310
  • Cas Number : 80914-40-5
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123624
Availability details for
Quantity
  • Availability
  • Add to Cart