ENA388700133 Enamine

3-[3-(methoxymethyl)-1,2,4-oxadiazol-5-yl]-2-methylaniline

  • MDL number MFCD16875113
  • Cas Number : 1239746-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123722

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123722
Availability details for
Quantity
  • Availability
  • Add to Cart