ENA388700138 Enamine

3-amino-2-{[4-(dimethylamino)phenyl]methyl}propanoic acid dihydrochloride

  • MDL number MFCD23144123
  • Cas Number : 1427380-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123733
Availability details for
Quantity
  • Availability
  • Add to Cart