ENA388700145 Enamine

6-methoxy-3-methyl-2,3,4,9-tetrahydro-1H-carbazole

  • MDL number MFCD21219393
  • Cas Number : 13220-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123746
Availability details for
Quantity
  • Availability
  • Add to Cart