ENA388700146 Enamine

5-chloro-3-ethyl-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04117281
  • Cas Number : 143214-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123747
Availability details for
Quantity
  • Availability
  • Add to Cart