ENA388700147 Enamine

methyl 2-[(4-nitrophenyl)amino]propanoate

  • MDL number MFCD12189072
  • Cas Number : 1311383-63-7
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123748
Availability details for
Quantity
  • Availability
  • Add to Cart