ENA388700151 Enamine

N-butyl-1-carbamimidamidomethanimidamide hydrochloride

  • MDL number MFCD00045591
  • Cas Number : 1190-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123753

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123753
Availability details for
Quantity
  • Availability
  • Add to Cart