ENA388700154 Enamine

1-[6-(aminomethyl)-2-methylpyridin-3-yl]ethan-1-ol dihydrochloride

  • MDL number MFCD23144127
  • Cas Number : 1427380-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123762
Availability details for
Quantity
  • Availability
  • Add to Cart