ENA388700160 Enamine

tert-butyl N-[(6-fluoro-1,2,3,4-tetrahydroquinolin-3-yl)methyl]carbamate

  • MDL number MFCD23144006
  • Cas Number : 1427379-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120511
Availability details for
Quantity
  • Availability
  • Add to Cart