ENA388700162 Enamine

2-(4-chlorophenyl)-1-(pyridin-2-yl)ethan-1-one

  • MDL number MFCD12153262
  • Cas Number : 204993-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123808

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123808
Availability details for
Quantity
  • Availability
  • Add to Cart