ENA388700164 Enamine

2-(4-chlorobenzoyl)-4-methylmorpholine

  • MDL number MFCD21791466
  • Cas Number : 1427379-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123810
Availability details for
Quantity
  • Availability
  • Add to Cart