ENA388700179 Enamine

3-(2,4,6-trifluorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD21162222
  • Cas Number : 1427380-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123832

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123832
Availability details for
Quantity
  • Availability
  • Add to Cart