ENA388700200 Enamine

propan-2-yl 2-(4-formyl-3,5-dimethyl-1H-pyrazol-1-yl)acetate

  • MDL number MFCD16073184
  • Cas Number : 1250348-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123880
Availability details for
Quantity
  • Availability
  • Add to Cart