ENA388700203 Enamine

5-[2-(3-methylbutanamido)ethyl]thiophene-2-sulfonyl chloride

  • MDL number MFCD09803821
  • Cas Number : 1016743-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123884
Availability details for
Quantity
  • Availability
  • Add to Cart