ENA388700214 Enamine

4-(2-bromophenyl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD09746484
  • Cas Number : 1095231-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123900
Availability details for
Quantity
  • Availability
  • Add to Cart