ENA388700215 Enamine

2-{[(3,4-dichlorophenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD07368396
  • Cas Number : 40172-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123901

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123901
Availability details for
Quantity
  • Availability
  • Add to Cart