ENA388700219 Enamine

4-(4-methoxybenzoyl)oxane

  • MDL number MFCD17243716
  • Cas Number : 1341760-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123906
Availability details for
Quantity
  • Availability
  • Add to Cart