ENA388700222 Enamine

1,3-diethyl 2-[2-(2-methoxyethoxy)ethyl]propanedioate

  • MDL number MFCD22353178
  • Cas Number : 667871-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123912
Availability details for
Quantity
  • Availability
  • Add to Cart