ENA388700226 Enamine

3-benzoyloxolane

  • MDL number MFCD14610314
  • Cas Number : 1248356-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123917
Availability details for
Quantity
  • Availability
  • Add to Cart