ENA388700227 Enamine

1-[1-(2-chlorophenyl)cyclobutyl]methanamine

  • MDL number MFCD16169400
  • Cas Number : 1227418-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123918
Availability details for
Quantity
  • Availability
  • Add to Cart