ENA388700228 Enamine

3-{[2-(1H-indol-3-yl)-2-phenylethyl]sulfamoyl}benzoic acid

  • MDL number MFCD07846171
  • Cas Number : 877966-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123923
Availability details for
Quantity
  • Availability
  • Add to Cart