ENA388700232 Enamine

4-{[2-(2-chlorophenyl)-2-(1H-indol-3-yl)ethyl]sulfamoyl}benzoic acid

  • MDL number MFCD11729598
  • Cas Number : 1053106-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123929
Availability details for
Quantity
  • Availability
  • Add to Cart