ENA388700235 Enamine

3-(2-methyl-5-oxopyrrolidin-2-yl)propanoic acid

  • MDL number MFCD20642356
  • Cas Number : 60769-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123935

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123935
Availability details for
Quantity
  • Availability
  • Add to Cart